siRNA Details
| Gene Symbol: CRISP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001131.2 | NP_001122.2 | 8 | 1410 | ||
| NM_001205220.1 | NP_001192149.1 | 8 | 1421 | |||
| NM_170609.1 | NP_733758.1 | 7 | 1321 | |||
| GenBank mRNA | AK292497.1 | - | 8 | 1391 | ||
| BC131707.1 | - | 7 | 1292 | |||
| GQ472203.1 | - | 8 | 1404 | |||
| X95237.1 | - | 8 | 1404 | |||
| X95238.1 | - | 7 | 1315 | |||

CRISP1
Genomic Map