siRNA Details
| Gene Symbol: AFP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134.2 | NP_001125.1 | 15 | 1910 | ||
| GenBank mRNA | AK297669.1 | - | 15 | 1822 | ||
| AK297674.1 | - | 15 | 1763 | |||
| BC027881.1 | - | 15 | 1896 | 5208299 | ||
| V01514.1 | - | 15 | 1910 | |||

AFP
Genomic Map