siRNA Details
| Gene Symbol: DFNA5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127453.1 | NP_001120925.1 | 4 | 534 | ||
| NM_001127454.1 | NP_001120926.1 | 3 | 549 | |||
| NM_004403.2 | NP_004394.1 | 4 | 779 | |||
| XM_017011802.1 | XP_016867291.1 | 3 | 305 | |||
| GenBank mRNA | AF073308.1 | - | 4 | 503 | ||
| AK094714.1 | - | 6 | 1107 | |||
| BC125065.1 | - | 4 | 501 | 40117291 | ||

DFNA5
Genomic Map