siRNA Details
| Gene Symbol: ALCAM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001243280.1 | NP_001230209.1 | 2 | 804 | ||
| NM_001243281.1 | NP_001230210.1 | 2 | 804 | |||
| NM_001243283.1 | NP_001230212.1 | 2 | 804 | |||
| NM_001627.3 | NP_001618.2 | 2 | 804 | |||
| GenBank mRNA | AK054632.1 | - | 2 | 757 | ||
| AK300362.1 | - | 2 | 180 | |||
| AK301804.1 | - | 3 | 237 | |||
| AK316113.1 | - | 2 | 608 | |||
| AL833702.1 | - | 2 | 605 | |||
| AY644765.1 | - | 2 | 456 | |||
| BC057809.1 | - | 2 | 608 | |||
| BC137096.1 | - | 2 | 232 | 9021093 | ||
| BC137097.1 | - | 2 | 232 | 9021094 | ||
| DQ486139.1 | - | 2 | 170 | |||
| DQ486140.1 | - | 2 | 170 | |||
| L38608.1 | - | 2 | 171 | |||
| Y10183.1 | - | 2 | 105 | |||

ALCAM
Genomic Map