siRNA Details
| Gene Symbol: AQP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001317384.1 | NP_001304313.1 | 5 | 2581 | ||
| NM_001317387.1 | NP_001304316.1 | 4 | 2500 | |||
| NM_001650.5 | NP_001641.1 | 5 | 2581 | |||
| NM_004028.4 | NP_004019.1 | 4 | 2708 | |||
| XM_011525942.2 | XP_011524244.1 | 5 | 2623 | |||
| GenBank mRNA | AB209156.1 | - | 3 | 2114 | ||
| AL119338.1 | - | 1 | 485 | |||
| AV725241.1 | - | 1 | 317 | |||
| BI917845.1 | - | 1 | 110 | |||
| CD105892.1 | - | 1 | 498 | |||

AQP4
Genomic Map