siRNA Details
| Gene Symbol: TRIM23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001656.3 | NP_001647.1 | 11 | 3781 | ||
| NM_033227.2 | NP_150230.1 | 13 | 2296 | |||
| NM_033228.2 | NP_150231.1 | 13 | 2267 | |||
| GenBank mRNA | BC022510.1 | - | 11 | 3721 | 4811707 | |

TRIM23
Genomic Map