siRNA Details
| Gene Symbol: ADGRB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001702.2 | NP_001693.2 | 9 | 2080 | ||
| XM_011517199.2 | XP_011515501.1 | 10 | 2518 | |||
| XM_011517202.2 | XP_011515504.1 | 10 | 2518 | |||
| XM_017013691.1 | XP_016869180.1 | 10 | 2518 | |||
| XM_017013692.1 | XP_016869181.1 | 10 | 2518 | |||
| XM_017013693.1 | XP_016869182.1 | 10 | 2509 | |||
| XM_017013694.1 | XP_016869183.1 | 10 | 2518 | |||
| XM_017013695.1 | XP_016869184.1 | 10 | 2518 | |||
| XM_017013696.1 | XP_016869185.1 | 10 | 2518 | |||
| XM_017013697.1 | XP_016869186.1 | 9 | 2353 | |||
| GenBank mRNA | AB005297.1 | - | 9 | 2080 | ||
| AK123982.1 | - | 2 | 737 | |||
More Information
Set Membership: |
|

ADGRB1
Genomic Map