siRNA Details
| Gene Symbol: ADGRB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001294335.1 | NP_001281264.1 | 8 | 1636 | ||
| NM_001294336.1 | NP_001281265.1 | 8 | 1636 | |||
| XM_011541848.2 | XP_011540150.1 | 7 | 1590 | |||
| XM_011541849.2 | XP_011540151.1 | 6 | 1425 | |||
| XM_011541858.2 | XP_011540160.1 | 5 | 589 | |||
| XM_017001899.1 | XP_016857388.1 | 7 | 1720 | |||
| XM_017001900.1 | XP_016857389.1 | 6 | 1692 | |||
| XM_017001901.1 | XP_016857390.1 | 6 | 1692 | |||
| XM_017001902.1 | XP_016857391.1 | 6 | 1693 | |||
| XM_017001903.1 | XP_016857392.1 | 6 | 1693 | |||
| XM_017001904.1 | XP_016857393.1 | 6 | 1689 | |||
| XM_017001905.1 | XP_016857394.1 | 6 | 1691 | |||
| XM_017001906.1 | XP_016857395.1 | 6 | 1690 | |||
| XM_017001907.1 | XP_016857396.1 | 5 | 1524 | |||
| XM_017001908.1 | XP_016857397.1 | 5 | 1525 | |||
| XM_017001909.1 | XP_016857398.1 | 5 | 1526 | |||
| XM_017001910.1 | XP_016857399.1 | 5 | 1524 | |||
| XM_017001911.1 | XP_016857400.1 | 5 | 1525 | |||
| XM_017001912.1 | XP_016857401.1 | 4 | 1358 | |||
| GenBank mRNA | AB005298.1 | - | 8 | 1608 | ||
| AK294999.1 | - | 4 | 529 | |||
| AK296502.1 | - | 3 | 268 | |||
| AK307805.1 | - | 5 | 1295 | |||
| BC136533.1 | - | 6 | 1304 | 9020523 | ||
More Information
Set Membership: |
|

ADGRB2
Genomic Map