siRNA Details
| Gene Symbol: BMP7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001719.2 | NP_001710.1 | 7 | 2052 | ||
| GenBank mRNA | AK094784.1 | - | 3 | 792 | ||
| AK291186.1 | - | 7 | 1628 | |||
| X51801.1 | - | 7 | 1645 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

BMP7| Gene Symbol: BMP7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001719.2 | NP_001710.1 | 7 | 2052 | ||
| GenBank mRNA | AK094784.1 | - | 3 | 792 | ||
| AK291186.1 | - | 7 | 1628 | |||
| X51801.1 | - | 7 | 1645 | |||