siRNA Details
| Gene Symbol: C1S | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001734.3 | NP_001725.1 | 12 | 1594 | ||
| NM_201442.2 | NP_958850.1 | 12 | 1681 | |||
| XM_005253760.1 | XP_005253817.1 | 12 | 1573 | |||
| GenBank mRNA | AK025309.1 | - | 9 | 1069 | ||
| AK290295.1 | - | 12 | 1628 | |||
| AM886411.1 | - | 11 | 1316 | |||
| BC056903.1 | - | 12 | 1621 | 6199186 | ||
| J04080.1 | - | 12 | 1520 | |||
| M18767.1 | - | 12 | 1540 | |||

C1S
Genomic Map