siRNA Details
| Gene Symbol: EPS8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004447.5 | NP_004438.3 | 4 | 588 | ||
| XM_011520605.2 | XP_011518907.1 | 5 | 648 | |||
| GenBank mRNA | AK291777.1 | - | 4 | 393 | ||
| AK292931.1 | - | 4 | 245 | |||
| AK304860.1 | - | 4 | 588 | |||
| BC030010.2 | - | 4 | 400 | 4820933 | ||
| BI667617.1 | - | 463 | ||||
| U12535.1 | - | 4 | 360 | |||

EPS8
Genomic Map