siRNA Details
| Gene Symbol: DYRK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003583.3 | NP_003574.1 | 2 | 528 | ||
| NM_006482.2 | NP_006473.2 | 3 | 677 | |||
| XM_017020032.1 | XP_016875521.1 | 2 | 820 | |||
| GenBank mRNA | AK313937.1 | - | 3 | 689 | ||
| BC005809.2 | - | 2 | 482 | 3831977 | ||
| BC006375.2 | - | 4 | 488 | 4139392 | ||
| Y13493.1 | - | 3 | 435 | |||

DYRK2
Genomic Map