siRNA Details
| Gene Symbol: FARSA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004461.2 | NP_004452.1 | 3 | 374 | ||
| GenBank mRNA | AF042347.1 | - | 3 | 326 | ||
| AK092093.1 | - | 3 | 341 | |||
| AK130195.1 | - | 3 | 345 | |||
| AK304587.1 | - | 3 | 346 | |||
| BC006495.2 | - | 3 | 329 | 2823551 | ||
| BC043565.1 | - | 3 | 324 | |||
| BT007198.1 | - | 3 | 326 | |||
| CR456740.1 | - | 3 | 326 | |||
| D84471.1 | - | 3 | 329 | |||
| U07424.1 | - | 3 | 338 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_025648, AK044637, AK171872, BC013533, AK002577, and others.

FARSA
Genomic Map