siRNA Details
| Gene Symbol: ERLIN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100626.1 | NP_001094096.1 | 9 | 753 | ||
| NM_006459.3 | NP_006450.2 | 8 | 997 | |||
| XM_005269442.3 | XP_005269499.1 | 9 | 783 | |||
| XM_017015541.1 | XP_016871030.1 | 9 | 767 | |||
| GenBank mRNA | AF064093.1 | - | 9 | 729 | ||
| AK056574.1 | - | 8 | 3354 | |||
| AK127017.1 | - | 2 | 2957 | |||
| AK222480.1 | - | 9 | 725 | |||
| AK298427.1 | - | 8 | 643 | |||
| AK315580.1 | - | 9 | 725 | |||
| BC031791.1 | - | 8 | 666 | 4214940 | ||

ERLIN1
Genomic Map