siRNA Details
| Gene Symbol: ERLIN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100626.1 | NP_001094096.1 | 12 | 1566 | ||
| NM_006459.3 | NP_006450.2 | 11 | 1810 | |||
| XM_005269442.3 | XP_005269499.1 | 12 | 1596 | |||
| XM_017015541.1 | XP_016871030.1 | 12 | 1580 | |||
| GenBank mRNA | AK127017.1 | - | 5 | 3770 | ||
| BC031791.1 | - | 11 | 1477 | 4214940 | ||

ERLIN1
Genomic Map