siRNA Details
| Gene Symbol: RERE | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001042681.1 | NP_001036146.1 | 7 | 1414 | ||
| NM_012102.3 | NP_036234.3 | 8 | 1599 | |||
| XM_005263464.2 | XP_005263521.1 | 7 | 1353 | |||
| XM_011541510.1 | XP_011539812.1 | 6 | 1339 | |||
| XM_011541511.1 | XP_011539813.1 | 7 | 1465 | |||
| XM_017001358.1 | XP_016856847.1 | 7 | 1104 | |||
| XM_017001359.1 | XP_016856848.1 | 7 | 1609 | |||
| GenBank mRNA | AB007927.2 | - | 6 | 749 | ||
| AB036737.1 | - | 8 | 1425 | |||
| AK096094.1 | - | 7 | 1341 | |||

RERE
Genomic Map