siRNA Details
| Gene Symbol: ARL2BP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012106.3 | NP_036238.1 | 6 | 650 | ||
| XM_011522977.1 | XP_011521279.1 | 6 | 389 | |||
| GenBank mRNA | AF126062.1 | - | 6 | 509 | ||
| AK075050.1 | - | 6 | 650 | |||
| AK311018.1 | - | 5 | 391 | |||
| BC003087.1 | - | 6 | 522 | 3503251 | ||
| BC094878.1 | - | 495 | 6728613 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_024191, NM_024269, AK202572, AK006418, AK150513, and others.

ARL2BP
Genomic Map