siRNA Details
| Gene Symbol: HSPB8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014365.2 | NP_055180.1 | 3 | 1533 | ||
| GenBank mRNA | AF133207.1 | - | 3 | 1370 | ||
| AF191017.1 | - | 3 | 1533 | |||
| AF217987.1 | - | 3 | 1378 | |||
| AF250138.1 | - | 3 | 1018 | |||
| AL136936.1 | - | 3 | 1363 | |||
| BC002673.2 | - | 3 | 1501 | 3607117 | ||

HSPB8
Genomic Map