siRNA Details
| Gene Symbol: BAHD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001301132.1 | NP_001288061.1 | 3 | 1892 | ||
| NM_014952.4 | NP_055767.3 | 3 | 1768 | |||
| XM_011521366.2 | XP_011519668.2 | 3 | 1892 | |||
| XM_011521367.2 | XP_011519669.2 | 3 | 1894 | |||
| XM_011521368.2 | XP_011519670.2 | 3 | 1891 | |||
| XM_011521369.2 | XP_011519671.1 | 3 | 1746 | |||
| GenBank mRNA | AB023162.1 | - | 3 | 1892 | ||
| AL833923.1 | - | 2 | 860 | |||
| BC022782.2 | - | 3 | 1707 | 3903581 | ||

BAHD1
Genomic Map