siRNA Details
| Gene Symbol: BAHD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001301132.1 | NP_001288061.1 | 7 | 4214 | ||
| NM_014952.4 | NP_055767.3 | 7 | 4090 | |||
| NR_125372.1 | 5 | 2252 | ||||
| XM_011521366.2 | XP_011519668.2 | 7 | 4214 | |||
| XM_011521367.2 | XP_011519669.2 | 7 | 4216 | |||
| XM_011521368.2 | XP_011519670.2 | 7 | 4204 | |||
| XM_011521369.2 | XP_011519671.1 | 7 | 4068 | |||
| GenBank mRNA | AB023162.1 | - | 7 | 4214 | ||
| AL833923.1 | - | 6 | 3173 | |||
| BC022782.2 | - | 7 | 4029 | 3903581 | ||

BAHD1
Genomic Map