siRNA Details
| Gene Symbol: ZC3H3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015117.2 | NP_055932.2 | 2 | 422 | ||
| XM_006716536.3 | XP_006716599.2 | 4 | 811 | |||
| XM_011516943.2 | XP_011515245.1 | 2 | 537 | |||
| XM_011516944.2 | XP_011515246.1 | 2 | 537 | |||
| XM_017013248.1 | XP_016868737.1 | 3 | 1442 | |||
| XM_017013249.1 | XP_016868738.1 | 3 | 984 | |||
| GenBank mRNA | BC038670.1 | - | 2 | 405 | 5260824 | |
| BM148116.1 | - | 2 | 425 | |||
| D63484.1 | - | 2 | 382 | |||

ZC3H3
Genomic Map