siRNA Details
| Gene Symbol: KANK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001136191.2 | NP_001129663.1 | 6 | 1801 | ||
| NM_015493.6 | NP_056308.3 | 4 | 1543 | |||
| GenBank mRNA | AB040951.2 | - | 5 | 1753 | ||
| AB284125.1 | - | 7 | 1984 | |||
| AK002094.1 | - | 3 | 233 | |||
| AK023332.1 | - | 3 | 285 | |||
| AY639929.1 | - | 4 | 1464 | |||
| BC082762.1 | - | 6 | 1755 | 6497026 | ||
| BC098105.1 | - | 5 | 1553 | 40012030 | ||
| BC098286.1 | - | 5 | 1553 | 40012033 | ||
| BC098312.1 | - | 5 | 1553 | 40012032 | ||

KANK2
Genomic Map