siRNA Details
| Gene Symbol: HOXB7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004502.3 | NP_004493.3 | 2 | 778 | ||
| GenBank mRNA | AK223249.1 | - | 2 | 777 | ||
| AK290653.1 | - | 2 | 744 | |||
| BC015345.1 | - | 2 | 725 | 4413080 | ||
| BM923808.1 | - | 2 | 779 | |||
| BQ233410.1 | - | 1 | 96 | |||
| M16937.1 | - | 2 | 774 | |||

HOXB7
Genomic Map