siRNA Details
| Gene Symbol: ARRDC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025604.2 | NP_001020775.1 | 8 | 1859 | ||
| NM_001286826.1 | NP_001273755.1 | 8 | 1831 | |||
| NM_015683.1 | NP_056498.1 | 8 | 1834 | |||
| GenBank mRNA | AF131826.1 | - | 7 | 1366 | ||
| AF193051.1 | - | 8 | 1834 | |||
| AK000689.1 | - | 8 | 1829 | |||
| AK127940.1 | - | 8 | 1823 | |||
| AK128588.1 | - | 2 | 3081 | |||
| BC022516.1 | - | 8 | 1832 | 4817429 | ||

ARRDC2
Genomic Map