siRNA Details
| Gene Symbol: NOL7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001317724.1 | NP_001304653.1 | 3, 4 | 413 | ||
| NM_016167.4 | NP_057251.2 | 3, 4 | 413 | |||
| GenBank mRNA | AF172066.3 | - | 3, 4 | 413 | ||
| AK075301.1 | - | 3, 4 | 396 | |||
| AL050299.1 | - | 2, 3 | 113 | |||
| BC023517.2 | - | 3, 4 | 388 | 4298095 | ||
| BC062683.1 | - | 3, 4 | 409 | 6499123 | ||
| BU071935.1 | - | 3, 4 | 166 | |||

NOL7
Genomic Map