siRNA Details
| Gene Symbol: UBAC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016172.2 | NP_057256.2 | 5, 6 | 763 | ||
| GenBank mRNA | AF038200.1 | - | 4, 5 | 328 | ||
| AF068195.1 | - | 4, 5 | 390 | |||
| AF176796.1 | - | 5, 6 | 736 | |||
| BC004967.1 | - | 5, 6 | 763 | 3535682 | ||
| BC011822.1 | - | 5, 6 | 589 | 3688526 | ||

UBAC1
Genomic Map