siRNA Details
| Gene Symbol: ZCCHC8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017612.4 | NP_060082.2 | 11, 12 | 1370 | ||
| XM_006719491.2 | XP_006719554.1 | 8, 9 | 1055 | |||
| XM_011538555.2 | XP_011536857.1 | 10, 11 | 1912 | |||
| XM_017019606.1 | XP_016875095.1 | 11, 12 | 1290 | |||
| GenBank mRNA | AK027702.1 | - | 11, 12 | 1239 | ||
| AK074638.1 | - | 11, 12 | 1268 | |||
| AK291174.1 | - | 7, 8 | 705 | |||
| AL157433.1 | - | 9, 10 | 2779 | |||
| BC017704.2 | - | 4, 5 | 494 | |||
| BC065918.1 | - | 11, 12 | 1209 | 5752053 | ||

ZCCHC8
Genomic Map