siRNA Details
| Gene Symbol: ZCCHC8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017612.4 | NP_060082.2 | 14 | 2526 | ||
| XM_006719491.2 | XP_006719554.1 | 11 | 2211 | |||
| XM_011538555.2 | XP_011536857.1 | 13 | 3068 | |||
| XM_017019606.1 | XP_016875095.1 | 14 | 2446 | |||
| GenBank mRNA | AK026831.1 | - | 3 | 1395 | ||
| AK027702.1 | - | 14 | 2395 | |||
| AK074638.1 | - | 14 | 2424 | |||
| AK291174.1 | - | 10 | 1861 | |||
| AL157433.1 | - | 12 | 3935 | |||
| BC017704.2 | - | 7 | 1650 | |||
| BC065918.1 | - | 14 | 2365 | 5752053 | ||

ZCCHC8
Genomic Map