siRNA Details
| Gene Symbol: DCHS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001142552.1 | NP_001136024.1 | 3 | 2805 | ||
| NM_017639.3 | NP_060109.2 | 5 | 653 | |||
| XM_011532045.1 | XP_011530347.1 | 3 | 2858 | |||
| XM_017008320.1 | XP_016863809.1 | 3 | 2858 | |||
| GenBank mRNA | AK127704.1 | - | 3 | 2797 | ||
| AY354497.1 | - | 5 | 653 | |||
| BC140919.1 | - | 3 | 2483 | |||

DCHS2
Genomic Map