siRNA Details
| Gene Symbol: VPS13C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001018088.2 | NP_001018098.1 | 4 | 352 | ||
| NM_017684.4 | NP_060154.3 | 4 | 352 | |||
| NM_018080.3 | NP_060550.2 | 4 | 352 | |||
| NM_020821.2 | NP_065872.1 | 4 | 352 | |||
| XM_011521713.2 | XP_011520015.1 | 4 | 313 | |||
| XM_011521714.2 | XP_011520016.1 | 4 | 313 | |||
| GenBank mRNA | AB037842.1 | - | 4 | 335 | ||
| AJ608770.1 | - | 4 | 335 | |||
| AJ608771.1 | - | 4 | 335 | |||
| AJ626860.1 | - | 4 | 335 | |||
| AJ626861.1 | - | 4 | 335 | |||
| AK307518.1 | - | 4 | 277 | |||

VPS13C
Genomic Map