siRNA Details
| Gene Symbol: PQLC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001040125.1 | NP_001035214.1 | 4 | 1032 | ||
| NM_001040126.1 | NP_001035215.1 | 4 | 420 | |||
| NM_017765.2 | NP_060235.2 | 5 | 662 | |||
| NR_109848.1 | 5 | 662 | ||||
| XM_005245916.2 | XP_005245973.1 | 4 | 445 | |||
| XM_011541634.2 | XP_011539936.1 | 5 | 614 | |||
| GenBank mRNA | AK000327.1 | - | 5 | 583 | ||
| AK125580.1 | - | 4 | 950 | |||
| AK130171.1 | - | 5 | 580 | |||
| AK130175.1 | - | 5 | 582 | |||
| BC015324.1 | - | 4 | 325 | 4426038 | ||
| KU178659.1 | - | 2 | 115 | |||

PQLC2
Genomic Map