siRNA Details
| Gene Symbol: CHCHD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001317177.1 | NP_001304106.1 | 6 | 655 | ||
| NM_017812.3 | NP_060282.1 | 5 | 640 | |||
| NR_133671.1 | 6 | 730 | ||||
| GenBank mRNA | AK000427.1 | - | 5 | 556 | ||
| AK129533.1 | - | 5 | 521 | |||
| AK310236.1 | - | 6 | 642 | |||
| AL516679.3 | - | 6 | 552 | |||
| BC011596.2 | - | 5 | 562 | 3346346 | ||
| BC014839.1 | - | 5 | 564 | 2901125 | ||

CHCHD3
Genomic Map