siRNA Details
| Gene Symbol: CHCHD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001317177.1 | NP_001304106.1 | 7, 8 | 758 | ||
| NM_017812.3 | NP_060282.1 | 6, 7 | 743 | |||
| GenBank mRNA | AK000427.1 | - | 6, 7 | 659 | ||
| AK129533.1 | - | 6, 7 | 624 | |||
| AL516679.3 | - | 7, 8 | 655 | |||
| BC011596.2 | - | 6, 7 | 665 | 3346346 | ||
| BC014839.1 | - | 6, 7 | 667 | 2901125 | ||

CHCHD3
Genomic Map