siRNA Details
| Gene Symbol: COMMD8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017845.3 | NP_060315.1 | 4, 5 | 536 | ||
| GenBank mRNA | AF182421.1 | - | 4, 5 | 566 | ||
| AK000509.1 | - | 4, 5 | 556 | |||
| AY542163.1 | - | 4, 5 | 528 | |||
| BC008371.1 | - | 4, 5 | 596 | 4051246 | ||
| BC015145.1 | - | 4, 5 | 540 | 4052767 | ||
| BC019826.1 | - | 4, 5 | 562 | 4995315 | ||

COMMD8
Genomic Map