siRNA Details
| Gene Symbol: TMEM38B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018112.2 | NP_060582.1 | 6 | 2026 | ||
| XM_005252075.2 | XP_005252132.1 | 6 | 2473 | |||
| XM_005252076.2 | XP_005252133.1 | 7 | 2043 | |||
| XM_005252077.3 | XP_005252134.1 | 7 | 2046 | |||
| XM_011518829.1 | XP_011517131.1 | 7 | 2035 | |||
| XM_011518832.2 | XP_011517134.1 | 4 | 1746 | |||
| XM_011518833.2 | XP_011517135.1 | 7 | 2158 | |||
| GenBank mRNA | AK001355.1 | - | 6 | 2007 | ||

TMEM38B
Genomic Map