siRNA Details
| Gene Symbol: TTC17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001307943.1 | NP_001294872.1 | 1, 2 | 234 | ||
| NM_018259.5 | NP_060729.2 | 1, 2 | 234 | |||
| XM_005253016.1 | XP_005253073.1 | 1, 2 | 234 | |||
| XM_011520218.2 | XP_011518520.1 | 1, 2 | 234 | |||
| XM_017018010.1 | XP_016873499.1 | 1, 2 | 234 | |||
| GenBank mRNA | AK001752.1 | - | 1, 2 | 187 | ||
| BC017238.2 | - | 1, 2 | 183 | 3940012 | ||
| BC033000.1 | - | 1, 2 | 180 | 4830377 | ||
| DA106610.1 | - | 1, 2 | 234 | |||

TTC17
Genomic Map