siRNA Details
| Gene Symbol: PDXP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020315.4 | NP_064711.1 | 2 | 1027 | ||
| GenBank mRNA | AK057265.1 | - | 2 | 593 | ||
| BC000320.2 | - | 2 | 1026 | 2821743 | ||
| BC009756.2 | - | 2 | 616 | |||
| BC018844.2 | - | 2 | 995 | |||
| BC064922.1 | - | 2 | 1027 | 6141538 | ||

PDXP
Genomic Map