siRNA Details
| Gene Symbol: PDXP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020315.4 | NP_064711.1 | 2 | 1514 | ||
| GenBank mRNA | AK057265.1 | - | 2 | 1080 | ||
| BC000320.2 | - | 2 | 1513 | 2821743 | ||
| BC009756.2 | - | 2 | 1103 | |||
| BC018844.2 | - | 2 | 1482 | |||
| BC064922.1 | - | 2 | 1514 | 6141538 | ||

PDXP
Genomic Map