siRNA Details
| Gene Symbol: SPC25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020675.3 | NP_065726.1 | 6 | 625 | ||
| XM_011511516.2 | XP_011509818.1 | 6 | 634 | |||
| GenBank mRNA | AF225416.1 | - | 6 | 607 | ||
| AK291093.1 | - | 6 | 636 | |||
| AY456388.1 | - | 5 | 494 | |||
| BC022255.1 | - | 6 | 625 | 4738187 | ||
| CR457197.1 | - | 5 | 494 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: AK146727, AK003408, AK011241, CX222514, BC033605, and others.

SPC25
Genomic Map