siRNA Details
| Gene Symbol: ZNF574 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022752.5 | NP_073589.4 | 2 | 717 | ||
| XM_005259159.2 | XP_005259216.1 | 1 | 580 | |||
| XM_011527228.2 | XP_011525530.1 | 2 | 910 | |||
| XM_011527229.2 | XP_011525531.1 | 2 | 633 | |||
| XM_011527230.2 | XP_011525532.1 | 2 | 585 | |||
| XM_017027149.1 | XP_016882638.1 | 2 | 1053 | |||
| GenBank mRNA | AF447880.1 | - | 2 | 661 | ||
| AK027895.1 | - | 2 | 620 | |||
| AK131369.1 | - | 2 | 1053 | |||
| AK131418.1 | - | 2 | 1203 | |||
| BC001184.1 | - | 2 | 606 | 3502564 | ||

ZNF574
Genomic Map