siRNA Details
| Gene Symbol: FNDC4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022823.2 | NP_073734.1 | 7 | 1524 | ||
| XM_005264499.3 | XP_005264556.1 | 6 | 1443 | |||
| GenBank mRNA | AK026015.1 | - | 7 | 1484 | ||
| AY358136.1 | - | 7 | 1242 | |||
| BC032725.1 | - | 6 | 1155 | 5533159 | ||

FNDC4
Genomic Map