siRNA Details
| Gene Symbol: NDUFS5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001184979.1 | NP_001171908.1 | 3 | 495 | ||
| NM_004552.2 | NP_004543.1 | 3 | 499 | |||
| GenBank mRNA | AF020352.1 | - | 3 | 449 | ||
| AF047434.1 | - | 3 | 460 | |||
| AF086268.1 | - | 3 | 450 | |||
| BC001884.1 | - | 3 | 417 | 3544747 | ||
| BC093057.1 | - | 3 | 439 | 30382835 | ||

NDUFS5
Genomic Map