siRNA Details
| Gene Symbol: ZNF577 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135590.1 | NP_001129062.1 | 7 | 2579 | ||
| NM_032679.2 | NP_116068.2 | 7 | 2732 | |||
| GenBank mRNA | BC004992.1 | - | 7 | 2700 | 2905976 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZNF577| Gene Symbol: ZNF577 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135590.1 | NP_001129062.1 | 7 | 2579 | ||
| NM_032679.2 | NP_116068.2 | 7 | 2732 | |||
| GenBank mRNA | BC004992.1 | - | 7 | 2700 | 2905976 | |