siRNA Details
| Gene Symbol: CTBS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004388.2 | NP_004379.1 | 2, 3 | 389 | ||
| GenBank mRNA | AK075423.1 | - | 2, 3 | 653 | ||
| AK298703.1 | - | 2, 3 | 340 | |||
| AK314640.1 | - | 2, 3 | 355 | |||
| BC126333.1 | - | 2, 3 | 365 | 8992049 | ||
| BC126335.1 | - | 2, 3 | 365 | 8992051 | ||
| M95767.1 | - | 2, 3 | 324 | |||

CTBS
Genomic Map