siRNA Details
| Gene Symbol: DAG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001165928.3 | NP_001159400.2 | 6 | 4485 | ||
| NM_001177634.2 | NP_001171105.1 | 6 | 4428 | |||
| NM_001177635.2 | NP_001171106.1 | 4 | 4017 | |||
| NM_001177636.2 | NP_001171107.1 | 5 | 4406 | |||
| NM_001177637.2 | NP_001171108.1 | 4 | 4244 | |||
| NM_001177638.2 | NP_001171109.1 | 4 | 4275 | |||
| NM_001177639.2 | NP_001171110.1 | 3 | 3965 | |||
| NM_001177640.2 | NP_001171111.1 | 4 | 4181 | |||
| NM_001177641.2 | NP_001171112.1 | 4 | 4271 | |||
| NM_001177642.2 | NP_001171113.1 | 4 | 4176 | |||
| NM_001177643.2 | NP_001171114.1 | 3 | 4175 | |||
| NM_001177644.2 | NP_001171115.1 | 3 | 4264 | |||
| NM_004393.5 | NP_004384.4 | 3 | 4124 | |||
| GenBank mRNA | BC012740.2 | - | 3 | 4059 | 4099382 | |
| BC014616.2 | - | 3 | 4059 | 2964740 | ||
| L19711.1 | - | 3 | 4100 | |||

DAG1
Genomic Map