siRNA Details
| Gene Symbol: MYO18A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_078471.3 | NP_510880.2 | 14 | 2656 | ||
| NM_203318.1 | NP_976063.1 | 14 | 2656 | |||
| GenBank mRNA | AB177858.1 | - | 13 | 2475 | ||
| AB177860.1 | - | 14 | 1608 | |||
| AK302505.1 | - | 13 | 1409 | |||
| AK303522.1 | - | 13 | 1546 | |||
| AY703984.1 | - | 14 | 1608 | |||
| BC039612.1 | - | 14 | 2760 | 6045482 | ||
| CR933614.1 | - | 14 | 2631 | |||
| D86970.2 | - | 14 | 2679 | |||

MYO18A
Genomic Map