siRNA Details
| Gene Symbol: ZNF551 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001270938.1 | NP_001257867.1 | 3 | 2226 | ||
| NM_138347.4 | NP_612356.2 | 3 | 2348 | |||
| NR_073102.1 | 2 | 2224 | ||||
| GenBank mRNA | AK091273.1 | - | 4 | 1384 | ||
| BC005868.2 | - | 3 | 2280 | 3530863 | ||
| BX538151.1 | - | 2 | 2194 | |||

ZNF551
Genomic Map