siRNA Details
| Gene Symbol: C2orf15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001317992.1 | NP_001304921.1 | 5 | 1165 | ||
| NM_144706.3 | NP_653307.1 | 4 | 1069 | |||
| GenBank mRNA | BC021264.2 | - | 4 | 960 | 4542657 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

C2orf15| Gene Symbol: C2orf15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001317992.1 | NP_001304921.1 | 5 | 1165 | ||
| NM_144706.3 | NP_653307.1 | 4 | 1069 | |||
| GenBank mRNA | BC021264.2 | - | 4 | 960 | 4542657 | |