siRNA Details
| Gene Symbol: FBXL14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152441.2 | NP_689654.1 | 2 | 1379 | ||
| XM_017018875.1 | XP_016874364.1 | 2 | 1361 | |||
| GenBank mRNA | BC028132.1 | - | 2 | 1379 | 5205284 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

FBXL14| Gene Symbol: FBXL14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152441.2 | NP_689654.1 | 2 | 1379 | ||
| XM_017018875.1 | XP_016874364.1 | 2 | 1361 | |||
| GenBank mRNA | BC028132.1 | - | 2 | 1379 | 5205284 | |