siRNA Details
| Gene Symbol: ZNF560 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152476.2 | NP_689689.2 | 10 | 2562 | ||
| XM_011527696.2 | XP_011525998.1 | 10 | 2591 | |||
| XM_011527697.2 | XP_011525999.1 | 10 | 2617 | |||
| XM_017026327.1 | XP_016881816.1 | 10 | 2571 | |||
| XM_017026328.1 | XP_016881817.1 | 10 | 2571 | |||
| XM_017026329.1 | XP_016881818.1 | 10 | 2571 | |||
| XM_017026330.1 | XP_016881819.1 | 10 | 2518 | |||
| GenBank mRNA | AK056548.1 | - | 10 | 2562 | ||
| BC101040.1 | - | 10 | 2527 | 40007878 | ||
| BC101041.1 | - | 10 | 2527 | 40007880 | ||
| BC101042.2 | - | 10 | 2527 | 40007881 | ||

ZNF560
Genomic Map